Learn More
IL-10 Monoclonal Antibody (JES5-16E3), FITC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£62.75 - £154.00
Specifications
Antigen | IL-10 |
---|---|
Clone | JES5-16E3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15288349
|
Affymetrix eBioscience
11-7101-81 |
50 μg |
£80.75
50µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15298349
|
Affymetrix eBioscience
11-7101-82 |
100 μg |
£154.00
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15596586
|
Affymetrix eBioscience
11-7101-41 |
25 Tests |
£62.75
25 tests |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The JES5-16E3 antibody reacts with mouse interleukin-10 (IL-10). Mouse IL-10 is an ∽18 kDa factor also known as Cytokine Synthesis Inhibitory Factor (CSIF). In the mouse, Th2 cells, B1 cells, macrophages, and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells, and acts as a costimulatory signal for mast cells, developing thymocytes and the Th2 response. Applications Reported: The JES5-16E3 antibody has been reported for use in capture of mouse IL-10 by ELISA and ELISPOT, intracellular staining for flow cytometric analysis, IHC, and neutralization of IL-10 bioactivity. Applications Tested: This JES5-16E3 antibody has been pre-titrated and tested by intracellular staining for flow cytometric analysis. test size: has been pre-titrated and tested by intracellular staining and flow cytometric analysis of LPS-stimulated normal human peripheral blood cells. This can be used at test size: 5 μL (0.25 μg)/test per test. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.Specifications
IL-10 | |
Flow Cytometry | |
FITC | |
Rat | |
Mouse | |
CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF | |
IL10 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IL10 |
JES5-16E3 | |
Monoclonal | |
Liquid | |
RUO | |
P18893 | |
16153 | |
IgG2b κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.