missing translation for 'onlineSavingsMsg'
Learn More

IL-10 Antibody (JES3-12G8), Biotin, Novus Biologicals™

Product Code. 18408421 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408421 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408421 Supplier Novus Biologicals Supplier No. NBP227577B0.1ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IL-10 Monoclonal antibody specifically detects IL-10 in Human, Cynomolgus Monkey, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), ELISA (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-10
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone JES3-12G8
Conjugate Biotin
Formulation PBS
Gene Alias CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Host Species Rat
Immunogen COS-expressed human IL-10
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3586
Target Species Human, Cynomolgus Monkey, Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.