missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IL-1 RII Antibody Pair [HRP]

Product Code. 18214458 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18214458

Brand: Novus Biologicals™ NBP27944115PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-1 RII.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 9.375 pg/mL
Assay Sensitivity 9.38-600 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human IL-1 RII.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725
Gene ID (Entrez) 7850
Gene Symbol IL1R2
Host Species Mouse/Rabbit
Kit Contents
  • Detection Antibody: 0.5 mg/mL mouse anti-IL-1 RII monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.5 mg/mL of rabbit anti-IL-1 RII monoclonal antibody.
  • Standard: Each vial contains 24 ng of recombinant IL-1 RII.
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cancer, Phospho Specific, Signal Transduction
    Storage Requirements Storage is content dependent.
    Target IL-1 RII
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating 2. Detection Antibody: Dilute to working concentration of 1.0 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve usi ng 2-fold serial dilutions in sample dilution buffer, and a high standard of 600 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.