missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-1 RII Monoclonal antibody specifically detects IL-1 RII in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | IL-1 RII |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 20 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | beta, CD121b, CD121b antigen, IL1RBCD121 antigen-like family member B, IL-1R-beta, IL-1RT2, IL-1RT-2, interleukin 1 receptor, type II, Interleukin-1 receptor beta, MGC47725 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-1 RII (Met 1-Glu 343; Accession#: NP_004624.1). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?