missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 RAcP/IL-1 R3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76924
This item is not returnable.
View return policy
Description
IL-1 RAcP/IL-1 R3 Polyclonal specifically detects IL-1 RAcP/IL-1 R3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| IL-1 RAcP/IL-1 R3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| C3orf13IL-1RAcPIL1R3Interleukin-1 receptor 3, FLJ37788, IL-1 receptor accessory protein, IL-1R3, IL-1R-3, interleukin 1 receptor accessory protein, interleukin-1 receptor accessory protein, interleukin-1 receptor accessory protein beta | |
| Rabbit | |
| 63 kDa | |
| 0.1 mg | |
| Cytokine Research, Immunology | |
| 3556 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL | |
| AF029213 | |
| IL1RAP | |
| Antibody was raised against a peptide corresponding to 22 amino acids near the carboxy terminus of human IL-1RAcP. The immunogen is located within the last 50 amino acids of IL-1RAcP. Amino Acid Squence: KKSPRRSSSDEQGLSYSSLKNV | |
| Affinity Purified | |
| RUO | |
| Primary | |
| It has no cross activity to other member in the IL-1 receptor family. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction