missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IL-1 RAcP/IL-1 R3 Polyclonal antibody specifically detects IL-1 RAcP/IL-1 R3 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | IL-1 RAcP/IL-1 R3 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C3orf13IL-1RAcPIL1R3Interleukin-1 receptor 3, FLJ37788, IL-1 receptor accessory protein, IL-1R3, IL-1R-3, interleukin 1 receptor accessory protein, interleukin-1 receptor accessory protein, interleukin-1 receptor accessory protein beta |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a peptide corresponding to 22 amino acids near the carboxy terminus of human IL-1RAcP. The immunogen is located within the last 50 amino acids of IL-1RAcP. Amino Acid Squence: KKSPRRSSSDEQGLSYSSLKNV |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?