Learn More
Invitrogen™ IL-1 beta Porcine ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ ESIL1BX5
Additional Details : Weight : 0.33800kg
Description
The Porcine Interleukin 1 beta (IL-1β) ELISA quantitates Porc IL-1β in porcine serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Porc IL-1β. Principle of the method The Porcine IL-1β solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.Specifications
P26889 | |
6 pg/mL | |
IL-1 beta ELISA Kit, 96 Assays, Swine/Porcine Species, IL1B Gene Aliases, Cytokines and Receptors Protein Family, The sandwich ELISA Antibody Pair Detects Porcine IL-1 beta, For Protein Assays and Analysis, 2°C to 8°C Storage Temperature | |
Biotin | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
96 assays | |
Colorimetric Microplate Reader | |
IL1B | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 100 μL | |
IL1B | |
1 hr. 20 min. |
8.23 pg/mL to 6000 pg/mL | |
6 pg/mL to 6000 pg/mL | |
Solid-phase sandwich ELISA | |
Colorimetric | |
Cytokines & Receptors | |
The sandwich ELISA antibody pair detects Porcine IL-1 beta. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
397122 | |
<12% | |
Anti-porcine IL-1 beta precoated 96-well Strip Plate 1 Ea., lyophilized recombinant porcine IL-1 beta standard 2 vials, 20 x wash buffer 25 mL, biotinylated antibody reagent 2 vials, streptavidin-HRP reagent 200 uL, TMB substrate 12 mL stop solution, contain | |
5 x 96 Tests | |
Immunology | |
2°C to 8°C | |
Porcine | |
4 hr. 45 min. |