missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 beta/IL-1F2 Antibody [mFluor Violet 610 SE], Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals AF37471MFV610
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Polyclonal antibody specifically detects IL-1 beta/IL-1F2 in Canine samples. It is validated for Neutralization,CyTOF
Specifications
| IL-1 beta/IL-1F2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Sheep | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Canine | |
| Purified |
| Neutralization, CyTOF | |
| mFluor Violet 610 SE | |
| catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
| E. coli-derived recombinant canine IL-1β/IL-1F2, Asp114-Ser265, Accession # Q28292 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Cardiovascular Biology, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Lipid and Metabolism, Metastasis | |
| 3553 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction