missing translation for 'onlineSavingsMsg'
Learn More

IL-1 beta/IL-1F2 Antibody (gevokizumab) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30172826
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30172826 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30172826 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP328830JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

IL-1 beta/IL-1F2 Monoclonal antibody specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1 beta/IL-1F2
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone gevokizumab
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Host Species Human
Immunogen IL-1b
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Cardiovascular Biology, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Lipid and Metabolism, Metastasis
Primary or Secondary Primary
Gene ID (Entrez) 3553
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.