missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 beta/IL-1F2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£184.00 - £407.00
Specifications
| Antigen | IL-1 beta/IL-1F2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence reported in scientific literature (PMID 30225853), Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18528900
|
Novus Biologicals
NBP1-19775SS |
0.025 mg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18517510
|
Novus Biologicals
NBP1-19775 |
0.1 mg |
£407.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-1 beta/IL-1F2 Polyclonal antibody specifically detects IL-1 beta/IL-1F2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| IL-1 beta/IL-1F2 | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence reported in scientific literature (PMID 30225853), Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| P01584 | |
| 3553 | |
| IgG | |
| Immunogen affinity purified |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Cardiovascular Biology, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Lipid and Metabolism, Metastasis | |
| PBS | |
| catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
| A synthetic peptide made to an internal sequence of human IL-1 beta [UniProt P01584]. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title