missing translation for 'onlineSavingsMsg'
Learn More

IL-1 beta/IL-1F2 Antibody (615417) [DyLight 680], Novus Biologicals™

Product Code. 30736122 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736122 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736122 Supplier Novus Biologicals Supplier No. FAB6964Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-1 beta/IL-1F2 Monoclonal antibody specifically detects IL-1 beta/IL-1F2 in Human samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1 beta/IL-1F2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 615417
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
Host Species Mouse
Immunogen E. coli-derived recombinant human IL-1β/IL-1F2 Propeptide, Met1-Asp116, Accession # P01584
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Cardiovascular Biology, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Infections (Virus Bacteria and Parasites), Inflammation, Innate Immunity, Lipid and Metabolism, Metastasis
Primary or Secondary Primary
Gene ID (Entrez) 3553
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.