missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 beta/IL-1F2 Antibody (43N1G6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80791
This item is not returnable.
View return policy
Description
IL-1 beta/IL-1F2 Monoclonal specifically detects IL-1 beta/IL-1F2 in Human, Rat samples. It is validated for Western Blot.
Specifications
| IL-1 beta/IL-1F2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1-5ug/ml | |
| catabolin, IL-1, IL-1 beta, IL-1B, IL1-BETA, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Rat reactivity reported in scientific literature (PMID: 28077335). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot | |
| 43N1G6 | |
| Unconjugated | |
| PBS | |
| IL1B | |
| Full-length recombinant human IL-1beta protein was used as immunogen for this antibody. | |
| 0.1 mg | |
| Adaptive Immunity, Apoptosis, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity, Metastasis | |
| 3553 | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction