Learn More
IL-1 Alpha Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28475-1-AP-150UL
This item is not returnable.
View return policy
Description
The IL-1 Alpha antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human IL-1 Alpha. This antibody recognizes human antigen. The IL-1 Alpha antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IL-1 Alpha | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Western Blot | |
| Unconjugated | |
| 150 μL | |
| 3552 | |
| IgG |