missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-1 alpha propeptide Polyclonal Antibody
GREENER_CHOICE

Sheep Polyclonal Antibody

Brand:  Invitrogen™ PA5148806

Product Code. 30534034

  • £415.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.2 mg/mL in sterile PBS.

Produced by activated macrophages, IL-1 stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity. IL-1 proteins are involved in the inflammatory response, being identified as endogenous pyrogens, and are reported to stimulate the release of prostaglandin and collagenase from synovial cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL-1 alpha propeptide
Polyclonal
Unconjugated
Il1a
hematopoietin-1; IL1; IL-1 alpha; IL1A; IL-1A; IL1-ALPHA; IL1F1; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin-1 alpha; preinterleukin 1 alpha; pro-interleukin-1-alpha
Sheep
Antigen affinity chromatography
RUO
3552
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Flow Cytometry, Western Blot
0.2 mg/mL
PBS with trehalose and no preservative
P01583
Il1a
E. coli-derived recombinant human IL-1 alpha /IL-1F1, Ala2-Arg112.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.