Learn More
PeproTech IL-1 alpha Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
£177.00 - £2745.00
Specifications
| Antigen | IL-1 alpha |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17845663
|
PeproTech
500-P180GBT-25UG |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17855413
|
PeproTech
500-P180GBT-50UG |
50 μg |
£228.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17825833
|
Invitrogen
500-P180GBT-1MG |
2 x 500 μg | |||||||
|
30054546
|
PeproTech
500-P180GBT-500UG |
500 μg |
£1466.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: MAPHSFQNNL RYKLIRIVKQ EFIMNDSLNQ NIYVDMDRIH LKAASLNDLQ LEVKFDMYAY SSGGDSKYPV TLKVSNTQLF VSAQGEDKPV LLKEIPETPK LITGSETDLI FFWEKINSKN YFTSAAFPEL LIATKEQSQV HLARGLPSMI DFQIS. Preparation: Produced from sera of goats immunized with highly pure Recombinant Rat IL-1alpha. Anti-Rat IL-1alpha-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat IL-1alpha by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat IL-1alpha (500-P180G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat IL-1alpha. Western Blot: To detect Rat IL-1alpha by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat IL-1alpha is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P180GBT-1MG will be provided as 2 x 500 μg
IL-1 alpha (Interleukin-1 alpha) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 alpha is coded by the IL1A gene and signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 beta. IL1A is located on the q arm on chromosome 2 at position 13. IL-1 alpha plays an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses. IL-1 alpha activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 alpha is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. Further, IL-1 alpha is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury and induces apoptosis. The IL1A gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL1 alpha regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL-1 alpha can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of IL-1 alpha are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.Specifications
| IL-1 alpha | |
| ELISA, Western Blot | |
| Biotin | |
| Goat | |
| Rat | |
| P16598 | |
| 24493 | |
| E.coli-derived Recombinant Rat IL-1alpha. | |
| Antigen affinity chromatography | |
| Il1a |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| CTLA8; hematopoietin-1; IL 1 a; IL 1α; IL1; IL-1 alpha; IL17; IL1A; Il-1a; IL-1alpha; IL1-ALPHA; IL1F1; IL1α; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1a; interleukin 1-alpha; interleukin 1-alpha (AA 1 - 152); Interleukin1 alpha; interleukin-1 alpha; interleukin-1 alpha precursor; Interleukin-17A; preinterleukin 1 alpha; pre-interleukin-1 alpha; pro-interleukin-1-alpha; RP23-160G19.8 | |
| Il1a | |
| Primary | |
| -20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.