missing translation for 'onlineSavingsMsg'
Learn More

IL-1 alpha/IL-1F1 Antibody, Novus Biologicals™

Product Code. 18409700 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.25 mg
Unit Size:
0.25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409700 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409700 Supplier Novus Biologicals Supplier No. NBP1778820.25mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IL-1 alpha/IL-1F1 Polyclonal antibody specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1 alpha/IL-1F1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:10 to 1:1000, ELISA 1:1000-1:5000, Immunohistochemistry 1:100 to 1:200, Immunohistochemistry-Paraffin 1:10 to 1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
Gene Accession No. P01583, P01583, P01583, P01583
Gene Alias Hematopoietin-1, IL1, IL-1 alpha, IL-1A, IL1-ALPHA, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, preinterleukin 1 alpha, pro-interleukin-1-alpha
Host Species Rabbit
Immunogen IL-1 alpha/IL-1F1 antibody was prepared by repeated immunizations with recombinant human IL-1 alpha/IL-1F1 produced in E.coli. The MW of the recombinant IL-1 alpha/IL-1F1 was 17,000. This is the cleavage site generated by the IL-1b converting enzyme (ICE, capase-1). (Uniprot: P01583.1)
Purification Method Whole Antiserum
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 3552
Target Species Human
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.