missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKZF5 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | IKZF5 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IKZF5 Polyclonal specifically detects IKZF5 in Human samples. It is validated for Western Blot.Specifications
| IKZF5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp781B0249, FLJ22973, IKAROS family zinc finger 5 (Pegasus), Ikaros family zinc finger protein 5, PEGASUS, zinc finger protein Pegasus, zinc finger protein, subfamily 1A, 5, zinc finger protein, subfamily 1A, 5 (Pegasus), zinc finger transcription factor Pegasus, ZNFN1A5 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human IKZF5. Peptide sequence TQAVVSAVSASIPQSSSPTSPEPRPSHSQRNYSPVAGPSSEPSAHTSTPS | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 64376 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title