missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IKZF4 Polyclonal Antibody
Description
Additional Information: This antibody is expected to recognise reported isoforms NP_071910.3; NP_001338019.1; NP_001338020.1; NP_001338021.1. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000.
DNA-binding protein that binds to the 5'GGGAATRCC-3' Ikaros-binding sequence. Transcriptional repressor. Interacts with SPI1 and MITF to repress transcription of the CTSK and ACP5 promoters via recruitment of corepressors SIN3A and CTBP2. May be involved in the development of central and peripheral nervous systems.
Specifications
Specifications
| Antigen | IKZF4 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | IKZF4 |
| Gene Accession No. | Q8C208, Q9H2S9 |
| Gene Alias | A630026H08Rik; Eos; IKAROS family zinc finger 4; IKAROS family zinc finger 4 (Eos); ikaros family zinc finger protein 4; Ikzf4; KIAA1782; Zfpn1a4; Zinc finger protein Eos; zinc finger protein, subfamily 1A, 4; zinc finger protein, subfamily 1A, 4 (Eos); zinc finger transcription factor Eos; ZNFN1A4 |
| Gene Symbols | IKZF4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?