missing translation for 'onlineSavingsMsg'
Learn More

IKK-IN-1, MedChemExpress

Product Code. p-200100177
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16445898

Brand: Medchem Express HY13873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IKK-IN-1 is an inhibitor of IKK extracted from patent WO2002024679A1, compound example 18-13.

IKK-IN-1 is an inhibitor of IKK extracted from patent WO2002024679A1, compound example 18-13.

Specifications

Chemical Name or Material IKK-IN-1
CAS 406211-06-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H26ClN3O4
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (77.17 mM; Need ultrasonic)
SMILES O=C1NC2=C(CO1)C(C3CCNCC3)=CC(C(C(O)=CC=C4)=C4OCC5CC5)=N2.Cl
Molecular Weight (g/mol) 431.91
Formula Weight 431.91
Percent Purity 95.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.