missing translation for 'onlineSavingsMsg'
Learn More

IKK-IN-1, MedChemExpress

Product Code. 16445898
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16445898 10 mg 10mg
16441389 1 mg 1mg
16438537 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16445898 Supplier Medchem Express Supplier No. HY13873
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress IKK-IN-1 is an inhibitor of IKK extracted from patent WO2002024679A1, compound example 18-13.

IKK-IN-1 is an inhibitor of IKK extracted from patent WO2002024679A1, compound example 18-13.

Specifications

Chemical Name or Material IKK-IN-1
CAS 406211-06-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H26ClN3O4
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (77.17 mM; Need ultrasonic)
SMILES O=C1NC2=C(CO1)C(C3CCNCC3)=CC(C(C(O)=CC=C4)=C4OCC5CC5)=N2.Cl
Molecular Weight (g/mol) 431.91
Formula Weight 431.91
Percent Purity 95.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.