missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IKK gamma Polyclonal Antibody

Description
In Western blot applications, this antibody detects a band at ~52kDa. A suggested positive control is Hela cell lysate. PA5-19965 can be used with blocking peptide PEP-0090.
IKK gamma is the regulatory subunit of the inhibitor of the kappaB kinase (IKK) complex, which activates NF-kappaB resulting in activation of genes involved in inflammation, immunity, cell survival, and other pathways. Mutations in this gene result in incontinentia pigmenti, hypohidrotic ectodermal dysplasia, and several other types of immunodeficiencies. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene highly similar to this locus is located in an adjacent region of the X chromosome.
Specifications
Specifications
| Antigen | IKK gamma |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Ikbkg |
| Gene Accession No. | O88522, Q6TMG5, Q9Y6K9 |
| Gene Alias | 1110037D23Rik; AI848108; AI851264; AMCBX1; AW124339; FIP3; FIP-3; Fip3p; gikk; I-kappa-B kinase gamma; I-kappa-B kinase subunit gamma; ikB kinase subunit gamma; ikB kinase-associated protein 1; IKBKG; IKK[g]; IKK3; IKKAP1; IKKG; IKK-gamma; IMD33; incontinentia pigmenti; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma; inhibitor of kappaB kinase gamma; inhibitor of nuclear factor kappa-B kinase subunit gamma; IP; IP1; IP2; IPD2; mF; mFIP-3; Nemo; NF-kappa B essential modulator; NF-kappa-B essential modifier; NF-kappa-B essential modulator; OTTHUMP00000207079; OTTHUMP00000212188; ZC2HC9 |
| Gene Symbols | Ikbkg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?