missing translation for 'onlineSavingsMsg'
Learn More

IKK gamma Antibody, mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18427322 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427322 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427322 Supplier Novus Biologicals Supplier No. NB600215MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IKK gamma Polyclonal antibody specifically detects IKK gamma in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKK gamma
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FIP-3IP2, FIP3NF-kappa-B essential modifier, Fip3p, I-kappa-B kinase subunit gamma, IkB kinase gamma subunit, IkB kinase subunit gamma, IkB kinase-associated protein 1, IKKAP1, IKKG, IKK-gammaIPD2, incontinentia pigmenti, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma, Inhibitor of nuclear factor kappa-B kinase subunit gamma, IP, IP1, NEMOAMCBX1, NFkappaB essential modulator, NF-kappa-B essential modulator
Host Species Rabbit
Immunogen N-terminus of the human IKK-gamma protein (proprietary).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Immunology, Innate Immunity, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 8517
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.