missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 7 Antibody (MM0433-5J28) - Azide and BSA Free, R&D Systems™

Product Code. 18038076 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18038076 0.025 mg 25µg
18095107 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18038076 Supplier Novus Biologicals Supplier No. NBP2117530.025MG
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Kallikrein 7 Monoclonal specifically detects Kallikrein 7 in Human samples. It is validated for Block/Neutralize, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 7
Applications Blocking Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone MM0433-5J28
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin 1:10-1:500, Blocking/Neutralizing 50-200 molar excess
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. NP_005037
Gene Alias EC 3.4.21, EC 3.4.21.117, hK7, hSCCE, kallikrein 7 (chymotryptic, stratum corneum), kallikrein-related peptidase 7, protease, serine, 6, PRSS6, SCCEkallikrein-7, Serine protease 6, signal protein, Stratum corneum chymotryptic enzyme
Gene Symbols KLK7
Host Species Mouse
Immunogen Human Kallikrein 7
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5650
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.