missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKK gamma Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
£150.00
Specifications
| Antigen | IKK gamma |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL, Flow (Intracellular) |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
IKK gamma Polyclonal specifically detects IKK gamma in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).Specifications
| IKK gamma | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 5 μg/mL, Flow (Intracellular) | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Immunology, Innate Immunity | |
| PBS with 0.02% Sodium Azide | |
| FIP-3IP2, FIP3NF-kappa-B essential modifier, Fip3p, I-kappa-B kinase subunit gamma, IkB kinase gamma subunit, IkB kinase subunit gamma, IkB kinase-associated protein 1, IKKAP1, IKKG, IKK-gammaIPD2, incontinentia pigmenti, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma, Inhibitor of nuclear factor kappa-B kinase subunit gamma, IP, IP1, NEMOAMCBX1, NFkappaB essential modulator, NF-kappa-B essential modulator | |
| IKBKG | |
| IgG | |
| Affinity Purified | |
| IKK gamma has no cross response to IKK alpha or IKK beta. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| AF074382 | |
| 8517 | |
| Antibody was raised against a 17 amino acid peptide near the carboxy terminus of human IKK gamma. The immunogen is located within the last 50 amino acids of IKK gamma. Amino Acid Squence: CQYQAPDMDTLQIHVME | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 52 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title