missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKK gamma Antibody (46B844), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
£177.00 - £397.00
Specifications
| Antigen | IKK gamma |
|---|---|
| Clone | 46B844 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2 ug/ml, Flow Cytometry 1:10-1:1000 |
| Applications | Western Blot, Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18036574
|
Novus Biologicals
NB100-56532SS |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18070274
|
Novus Biologicals
NB100-56532 |
0.1 mg |
£397.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IKK gamma Monoclonal specifically detects IKK gamma in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
| IKK gamma | |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Immunology, Innate Immunity | |
| NP_001093326 | |
| 8517 | |
| Full-length human IKK gamma protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 46B844 | |
| Western Blot 2 ug/ml, Flow Cytometry 1:10-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FIP-3IP2, FIP3NF-kappa-B essential modifier, Fip3p, I-kappa-B kinase subunit gamma, IkB kinase gamma subunit, IkB kinase subunit gamma, IkB kinase-associated protein 1, IKKAP1, IKKG, IKK-gammaIPD2, incontinentia pigmenti, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma, Inhibitor of nuclear factor kappa-B kinase subunit gamma, IP, IP1, NEMOAMCBX1, NFkappaB essential modulator, NF-kappa-B essential modulator | |
| IKBKG | |
| IgG1 | |
| Protein G purified | |
| A 48 kDa band is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title