Learn More
Invitrogen™ IKK epsilon Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587387
Description
Positive Samples: SW480, DU145, MCF7, Mouse liver; Cellular Location: Cytoplasm, Nucleus, PML body Immunogen sequence: SRLRTLAEVL SRCSQNITET QESLSSLNRE LVKSRDQVHE DRSIQQIQCC LDKMNFIYKQ FKKSRMRPGL GYNEEQIHKL DKVNFSHLAK RLLQVFQEEC VQKYQASLVT HGKRMRVVHE TRNHLRLVGC SVAACNTEAQ GVQESLSKLL EELSHQLLQD RAKGAQASPP PIAPYPSPTR KDLLLHMQEL CEGMKLLASD LLDNNRIIER LNRVPAPPDV.
IKBKE is an activator of nuclear factor-kappa-B. Over expression of IKBKE has been observed in breast cancer cells, and suppression of IKBKE induces cell death. NF-kappa-B is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kappa-B mediates the expression of a great variety of genes in response to extracellular stimuli. NF-kappa-B is associated with Ikappa-B proteins in the cell cytoplasm, which inhibit NF-kappa-B activity. Ikappa-B is phosphorylated by Ikappa-B kinase (IKK) complex that contains IKK-a, IKK-b, and IKK-gamma. A novel molecule in the IKK complex was recently identified and designated IKK-iota/IKK-i. IKK epsilon is required for the activation of NF-kappa-B by PMA and T cell receptors but not by TNF-a and IL-1. IKK-iota/IKK-i message is expressed in a variety of tissues and is inducible by TNF-a, IL-1, and LPS.
Specifications
| IKK epsilon | |
| Polyclonal | |
| Unconjugated | |
| IKBKE | |
| AW558201; I-kappa-B kinase epsilon; Ikbke; IKKE; IKK-E; IKKepsilon; IKK-epsilon; IKKI; IKK-i; IKK-related kinase epsilon; inducible I kappa-B kinase; inducible IkappaB kinase; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon; inhibitor of kappaB kinase epsilon; inhibitor of nuclear factor kappa-B kinase subunit epsilon; KIAA0151; RP11-534L20.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 363984, 56489, 9641 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.58 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q14164, Q9R0T8 | |
| IKBKE | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 497-716 of human IKK? (NP_0547211). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.