Learn More
IKK epsilon/IKBKE, p Thr501 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand:  Novus Biologicals NBP1-77829
Product Code. 18708343
  • £536.00 / 0.10mg
Description
IKK epsilon/IKBKE Polyclonal specifically detects IKK epsilon/IKBKE in Human samples. It is validated for Western Blot, Simple Western, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Affinity Purified
IgG
1 mg/mL
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.