Learn More
Invitrogen™ IKK beta Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5120783
Product Code. 17712184
  • £437.00 / 100µL
Description
Positive test controls include: HeLa, A-549, Jurkat, HepG2, THP-1. The target is usually found in the following locations: Cytoplasm, Membrane raft, Nucleus. Immunogen sequence: QISPRPQPES VSCILQEPKR NLAFFQLRKV WGQVWHSIQT LKEDCNRLQQ GQRAAMMNLL RNNSCLSKMK NSMASMSQQL KAKLDFFKTS IQIDLEKYSE QTEFGITSDK LLLAWREMEQ AVELCGRENE VKLLVERMMA LQTDIVDLQR SPMGRKQGGT LDDLEEQARE LYRRLREKPR DQRTEGDSQE MVRLLLQAIQ SFEKKVRVIY TQLSKTVVCK QKALELLPKV EEVVSLMNED EKTVVRLQEK RQKELWNLLK IACSKVRGPV.
Phosphorylation of serine residues on the I-kappa-B proteins by IKK kinases, including IKBKB, marks them for destruction via the ubiquitination pathway, allowing activation of the NF-kappa-B complex. Inhibitor of nuclear factor kappa-B kinase subunit beta (IKK beta) is a serine kinase that plays an essential role in the NF-kaapa-B signaling pathway which is activated by mulitple stimuli such as inflammatory cytokines, bacterial, or viral products, DNA damages, or other cellular stresses. IKK beta acts as part of the canonical IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B on 2 critical serine residues. These modifications allow polyubiquitination of the inhibitors and subsequent degradation by the proteasome. In turn, free NF-kappa-B is translocated into the nucleus and activates the transcription of hundreds of genes involved in immune response, growth control, or protection against apoptosis. Mutations affecting the IKK beta gene can result in Immunodeficiency 15 (IMD15).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IKK beta
Polyclonal
Unconjugated
Ikbkb
AI132552; AIM-1; FLJ40509; I-kappa-B kinase 2; I-kappa-B-kinase beta; Ikbkb; IKK[b]; IKK2; IKK-2; IKKB; IKK-B; IKKbeta; IKK-beta; IMD15; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta; inhibitor of kappaB kinase beta; inhibitor of nuclear factor kappa B kinase beta subunit; inhibitor of nuclear factor kappa-B kinase subunit beta; MGC131801; NFKBIKB; nuclear factor NF-kappa-B inhibitor kinase beta
Rabbit
Affinity chromatography
RUO
16150, 3551, 84351
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
10.45 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
O14920, O88351, Q9QY78
Ikbkb
A synthetic peptide corresponding to a sequence within amino acids 400-669 of human IKK beta (NP_0015471).
100 μL
Primary
Human, Mouse, Rat