missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKBKAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | IKBKAP |
|---|---|
| Dilution | Western Blot 1:1000 - 1:4000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30230350
|
Novus Biologicals
NBP3-35099-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230383
|
Novus Biologicals
NBP3-35099-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IKBKAP Polyclonal antibody specifically detects IKBKAP in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| IKBKAP | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Transcription Factors and Regulators | |
| PBS (pH 7.3), 50% glycerol | |
| 8518 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:4000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DYS, elongator complex protein 1, ELP1DKFZp781H1425, FD, FLJ12497, IKAPdysautonomia (Riley-Day syndrome, hereditary sensory autonomic neuropathy typeIII), IkappaB kinase complex-associated protein, IKI3, IKK complex-associated protein, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinasecomplex-associated protein, p150, TOT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1093-1332 of human IKBKAP (NP_003631.2).,, Sequence:, LRLVYKYNRLDIIETNVKPSILEAQKNYMAFLDSQTATFSRHKKRLLVVRELKEQAQQAGLDDEVPHGQESDLFSETSSVVSGSEMSGKYSHSNSRISARSSKNRRKAERKKHSLKEGSPLEDLALLEALSEVVQNTENLKDEVYHILKVLFLFEFDEQGRELQKAFEDTLQLMERSLPEIWTLTYQQNSATPVLGPNSTANSIMASYQQQKTSVPVLDAELFIPPKINRRTQWKLSLLD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title