missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
IKBKAP Polyclonal antibody specifically detects IKBKAP in Human samples. It is validated for Western Blot
Specifications
Specifications
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Amino acid residues 148-161 IHQDDFGESKFITV of human IKAP. Cross reactivity: sequence 92% identical to rat and 85% identical to mouse. |
| Regulatory Status | RUO |
| Target Species | Human |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?