missing translation for 'onlineSavingsMsg'
Learn More

IKBKAP Antibody, DyLight 594, Novus Biologicals™

Product Code. 18418214 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418214 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418214 Supplier Novus Biologicals Supplier No. NBP171918DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IKBKAP Polyclonal antibody specifically detects IKBKAP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen IKBKAP
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 594
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DYS, elongator complex protein 1, ELP1DKFZp781H1425, FD, FLJ12497, IKAPdysautonomia (Riley-Day syndrome, hereditary sensory autonomic neuropathy typeIII), IkappaB kinase complex-associated protein, IKI3, IKK complex-associated protein, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinasecomplex-associated protein, p150, TOT1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1150 and 1200 of human Inhibitor of Kappa Light Polypeptide Gene Enhancer in B-cells, Kiinase Complex-Associated Protein using the numbering given in entry NP_003631.2 (GeneID 8518).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 8518
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.