missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKBKAP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00
Specifications
| Antigen | IKBKAP |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1-20 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
IKBKAP Polyclonal specifically detects IKBKAP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.Specifications
| IKBKAP | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1-20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Cancer, Transcription Factors and Regulators | |
| PBS with 0.02% Sodium Azide | |
| DYS, elongator complex protein 1, ELP1DKFZp781H1425, FD, FLJ12497, IKAPdysautonomia (Riley-Day syndrome, hereditary sensory autonomic neuropathy typeIII), IkappaB kinase complex-associated protein, IKI3, IKK complex-associated protein, inhibitor of kappa light polypeptide gene enhancer in B-cells, kinasecomplex-associated protein, p150, TOT1 | |
| ELP1 | |
| IgG | |
| Affinity Purified | |
| At least two isoforms of IKAP are known two exist, this antibody will detect both isoforms. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| AAC64258 | |
| 8518 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human IKAP. The immunogen is located within the last 50 amino acids of IKAP. Amino Acid Squence: PPKINRRTQWKLSLLD | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 105, 145 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title