missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IkB zeta Recombinant Rat Monoclonal Antibody (LK2NAP)

Rat Recombinant Monoclonal Antibody

Brand:  Invitrogen™ 740065T

 View more versions of this product

Product Code. 30276570

  • £383.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recombinant rat monoclonal antibodies are produced using in vitro expression systems. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include better specificity and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

I-kappa-B-zeta (also known as MAIL and INAP) is an ankyrin-repeat-containing nuclear protein that is highly homologous to the IkappaB family member Bcl-3. Transcription of IkappaBzeta is upregulated by stimulation with TLR ligands, IL-1 and IL-6 in cultured B-lymphocytes and monocytes/macrophages, but only faintly so in T-lymphocytes, fibroblasts, and endothelial cells. IkappaB-zeta preferentially associates with the NF-kappaB subunit p50 rather than p65 and recombinant IkappaB-zeta proteins inhibit the DNA binding of the p65/p50 heterodimer and the p50/p50 homodimer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IkB zeta
Recombinant Monoclonal
1.0 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9BYH8, Q9EST8
NFKBIZ
Affinity chromatography
RUO
304005, 64332, 80859
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
LK2NAP
Unconjugated
NFKBIZ
AA408868; FLJ30225; FLJ34463; Ikappa B-zeta variant 3; ikappaBzeta; I-kappa-B-zeta; IkappaB-zeta; IKBZ; ikB-zeta; IL-1 inducible nuclear ankyrin-repeat protein; INAP; Mail; molecule possessing ankyrin repeats induced by lipopolysaccharide; molecule possessing ankyrin-repeats induced by lipopolysaccharide; NF-kappa-B inhibitor zeta; NFKB inhibitor zeta; NFKBIZ; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, zeta; OTTHUMP00000214340; RGD1310834
Rat
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.