missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IkB zeta Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567382
This item is not returnable.
View return policy
Description
Immunogen sequence: CQPFQVRGSQQ MIDQASLYQY SPQNQHVEQQ PHYTHKPTLE YSPFPIPPQS PAYEPNLFDG PESQFCPNQS LVSLLGDQRE SENIANPM Highest antigen sequence identity to the following orthologs - mouse 49%, rat 39%.
I-kappa-B-zeta (also known as MAIL and INAP) is an ankyrin-repeat-containing nuclear protein that is highly homologous to the IkappaB family member Bcl-3. Transcription of IkappaBzeta is upregulated by stimulation with TLR ligands, IL-1 and IL-6 in cultured B-lymphocytes and monocytes/macrophages, but only faintly so in T-lymphocytes, fibroblasts, and endothelial cells. IkappaB-zeta preferentially associates with the NF-kappaB subunit p50 rather than p65 and recombinant IkappaB-zeta proteins inhibit the DNA binding of the p65/p50 heterodimer and the p50/p50 homodimer.
Specifications
| IkB zeta | |
| Polyclonal | |
| Unconjugated | |
| NFKBIZ | |
| AA408868; FLJ30225; FLJ34463; Ikappa B-zeta variant 3; ikappaBzeta; I-kappa-B-zeta; IkappaB-zeta; IKBZ; ikB-zeta; IL-1 inducible nuclear ankyrin-repeat protein; INAP; Mail; molecule possessing ankyrin repeats induced by lipopolysaccharide; molecule possessing ankyrin-repeats induced by lipopolysaccharide; NF-kappa-B inhibitor zeta; NFKB inhibitor zeta; NFKBIZ; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, zeta; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, zeta; OTTHUMP00000214340; RGD1310834 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64332 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BYH8 | |
| NFKBIZ | |
| Recombinant Human IkB zeta. Recombinant protein control fragment (Product #RP-108043). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction