missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IkB epsilon Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582029
This item is not returnable.
View return policy
Description
Immunogen sequence: PEPGRGTSHS LDLQLQNWQG LACLHIATLQ KNQPLMELLL RNGADIDVQE GTSGKTALHL AVETQERGLV QFLLQAGAQV DARMLNGCTP LHLAAGRGLM GISSTLCKAG ADSLLRNVED ETPQDLTEES LVLLPFDDLK I Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 87%.
NFKB1 or NFKB2 is bound to REL, RELA, or RELB to form the NFKB complex. The NFKB complex is inhibited by I-kappa-B proteins (NFKBIA, MIM 164008 or NFKBIB, MIM 604495), which inactivate NF-kappa-B by trapping it in the cytoplasm. Phosphorylation of serine residues on the I-kappa-B proteins by kinases (IKBKA, MIM 600664, or IKBKB, MIM 603258) marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B complex. Activated NFKB complex translocates into the nucleus and binds DNA at kappa-B-binding motifs such as 5-prime GGGRNNYYCC 3-prime or 5-prime HGGARNYYCC 3-prime (where H is A, C, or T; R is an A or G purine; and Y is a C or T pyrimidine). For some genes, activation requires NFKB interaction with other transcription factors, such as STAT (see STAT6, MIM 601512), AP1 (JUN, MIM 165160), and NFAT (see NFATC1, MIM 600489).
Specifications
| IkB epsilon | |
| Polyclonal | |
| Unconjugated | |
| Nfkbie | |
| ikappaBepsilon; I-kappa-B-epsilon; IkB epsilon; IKBE; IkB-E; IkBepsilon; ikB-epsilon; NF-kappa-B inhibitor epsilon; NF-kappa-BIE; NFKB inhibitor epsilon; NFKBIE; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, epsilon; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, epsilon; Slc35b2; solute carrier family 35, member B2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4794 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00221 | |
| Nfkbie | |
| Recombinant protein corresponding to Human NFKBIE. Recombinant protein control fragment (Product #RP-99393). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction