missing translation for 'onlineSavingsMsg'
Learn More

IkB-beta, p Ser23 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18412844 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412844 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412844 Supplier Novus Biologicals Supplier No. NBP254761FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IkB-beta Polyclonal antibody specifically detects IkB-beta in Human, Mouse, Rat samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen IkB-beta
Applications Immunohistochemistry
Classification Polyclonal
Conjugate DyLight 680
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias beta, IkappaBbeta, I-kappa-B-beta, IkB-B, IkB-beta, IKBBikB-beta, NF-kappa-B inhibitor beta, NF-kappa-BIB, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, Thyroid receptor-interacting protein 9, TR-interacting protein 9, TRIP-9, TRIP9ikappaBbeta
Host Species Rabbit
Immunogen IkB-beta (Phospho-Ser23) antibody was raised against a peptide sequence around phosphorylation site of serine 23 (L-G-S (p) -L-G) derived from Human IkappaB-b
Purification Method Epitope affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4793
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.