missing translation for 'onlineSavingsMsg'
Learn More

IkB-beta Antibody (2B11), Novus Biologicals™

Product Code. 18328898 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328898 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328898 Supplier Novus Biologicals Supplier No. H00004793M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IkB-beta Monoclonal antibody specifically detects IkB-beta in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen IkB-beta
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2B11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH15528
Gene Alias beta, IkappaBbeta, I-kappa-B-beta, IkB-B, IkB-beta, IKBBikB-beta, NF-kappa-B inhibitor beta, NF-kappa-BIB, nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, Thyroid receptor-interacting protein 9, TR-interacting protein 9, TRIP-9, TRIP9ikappaBbeta
Host Species Mouse
Immunogen NFKBIB (AAH15528, 56 a.a. ~ 145 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EDGDTALHLAVIHQHEPFLDFLLGFSAGTEYMDLQNDLGQTALHLAAILGETSTVEKLYAAGAGLCVAERRGHTALHLACRVGAHACARA
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 4793
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.