Learn More
Invitrogen™ IkB alpha Recombinant Rabbit Monoclonal Antibody (1I1F7)

Description
Immunogen sequence: MFQPAGHGQD WAMEGPRDGL KKERLVDDRH DSGLDSMKDE DYEQMVKELR EIRLQPQEAP LAAEPWKQQL TEDGDSFLHL AIIHEEKTLT MEVIGQVKGD LAFLNFQNNL QQTPLHLAVI TNQPGIAEAL LKAGCDPELR DFRGNTPLHL ACEQGCLASV AVLTQTCTPQ HLHSVLQATN YNGHTCLHLA SIHGYLGIVE HLVTLGADVN AQEPCNGRTA LHLAVDLQNP DLVSLLLKCG ADVNRVTYQG YSPYQLTWGR PSTRIQQQLG QLTLENLQTL PESEDEESYD TESEFTEDEL PYDDCVFGGQ RLTL.
Specifications
Specifications
| Antigen | IkB alpha |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 1I1F7 |
| Concentration | 1.73 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | NFKBIA |
| Gene Accession No. | P25963, Q63746, Q9Z1E3 |
| Gene Alias | AI462015; ECI-6; ECI-6/IKBA; I kappa B-alpha; I(Kappa)B(alpha); I79_018146; IkappaB alpha; IkappaBalpha; I-kappaBalpha; I-kappa-B-alpha; Ikba; IKBalpha; IKB-alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells alpha; Inhibitor of nuclear factor of kappa light chain gene enhancer in B-cells, alpha; MAD3; MAD-3; Major histocompatibility complex enhancer-binding protein MAD3; NF kappa B inhibitor alpha; NF-kappaB inhibitor alpha; NF-kappa-B inhibitor alpha; NFKB inhibitor alpha; NFKBI; Nfkbia; nuclear factor of kappa light chain gene enhancer in B-cells; nuclear factor of kappa light chain gene enhancer in B-cells inhibitor, alpha; nuclear factor of kappa light polyp gene enhancer in B-cell 1; nuclear factor of kappa light polypeptide gene enhancer in B cells inhibitor, alpha; nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha; Rel-associated pp40; REL-associated protein pp40; RL/IF-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.