missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IKAP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111320
This item is not returnable.
View return policy
Description
Immunogen sequence: RLHVLCQGWH YLAYDWHWTT DRSVGDNSSD LSNVAVIDGN RVLVTVFRQT VVPPPMCTYQ LLFPHPVNQV TFLAHPQKSN DLAVLDASNQ ISVYKCGDCP SADPTVKLGA VG Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 77%.
The protein encoded by this gene is a scaffold protein and a regulator for 3 different kinases involved in proinflammatory signaling. This encoded protein can bind NF-kappa-B-inducing kinase (NIK) and IKKs through separate domains and assemble them into an active kinase complex. Mutations in this gene have been associated with familial dysautonomia.
Specifications
| IKAP | |
| Polyclonal | |
| Unconjugated | |
| ELP1 | |
| 3110040G09Rik; 6030413P05; C78473; DKFZp781H1425; DYS; elongator acetyltransferase complex subunit 1; elongator complex protein 1; Elp1; FD; FLJ12497; IKAP; IkappaB kinase complex-associated protein; IKBKAP; IKI3; IKK complex-associated protein; IKK-complex-associated protein IKAP; inhibitor of kappa light polypeptide enhancer in B cells, kinase complex-associated protein; inhibitor of kappa light polypeptide enhancer in B-cells, kinase complex-associated protein; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein; p150; RP11-3J11.4; TOT1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8518 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95163 | |
| ELP1 | |
| Recombinant Protein corresonding to Human IKAP. Recombinant protein control fragment (Product #RP-103837). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction