Learn More
IKA RV 10.4 Condensate Trap
Shop All IKA Products
Click to view available options
For Use With (Equipment):
For Distilling Low-Boiling Solvents
Unit Size:
Each
Description
- The solvent to be distilled can be continuously fed in through the PTFE inlet tube
- Cooling by dry ice, no cooling water required
- Max. condensation thanks to low temperatures
Specifications
Specifications
| Type | Dry Ice Condenser |
| For Use With (Application) | For Distilling Low-Boiling Solvents |
| For Use With (Equipment) | For Distilling Low-Boiling Solvents |