missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IKA™ MS 3 Digital Shaker

Product Code. 31784131 Shop All Applied Biosystems Products
Change view
Click to view available options
Speed Range:
0 to 3000 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range unitSize
31784131 0 to 3000 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31784131 Supplier Invitrogen™ Supplier No. QP0706
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The IKA MS 3 Digital Shaker is a compact, universal shaker suitable for room temperature shaking tasks in the QuantiGene Plex assay protocol.

The IKA MS 3 Digital Shaker is a compact, universal shaker suitable for room temperature shaking tasks in the QuantiGene Plex assay protocol.

The IKA MS 3 digital shaker has the following features:

  • Versatile configuration—a wide range of attachments and attachment detection
  • Precision—includes a timer with countdown function
  • Ease of operation—has a continuous or touch operation (with standard attachment)
  • Flexible operating modes:
    • Mode A (safe mode with attachment detection)—the maximum speed of 3,000 rpm is only reached with the standard attachment in touch mode. When using other attachments, the speed is limited to 1,300 rpm.
    • Mode B (without attachment detection)—a speed of 3,000 rpm is possible with all attachments
  • Stable action in all speed ranges
  • Sturdy construction—zinc die cast casing
TRUSTED_SUSTAINABILITY

Specifications

Speed Range 0 to 3000 rpm
Product Type Digital Shaker
Certifications/Compliance Certificate of Analysis (COA)
Model IKA MS 3 Digital Shaker
Display Digital
Product Line IKA
Quantity 1 shaker

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.