missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IHH Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA579455
This item is not returnable.
View return policy
Description
The synthetic peptide sequence is 250-264aa, EPHRLRAFQVIETQD Add 0.2 mL of distilled water, will yield a concentration of 500 μg/mL.
This gene encodes a member of the hedgehog family of secreted signaling molecules. Hedgehog proteins are essential regulators of a variety of developmental processes including growth, patterning and morphogenesis. The encoded protein specifically plays a role in bone growth an differentiation. Mutations in this gene are the cause of brachydactyly type A1 which is characterized by shortening or malformation of the phalanges. Mutations in this gene are also the cause of acrocapitofemoral dysplasia.
Specifications
| IHH | |
| Polyclonal | |
| PBS with 5MG BSA, 5MG BSA and 0.05MG sodium azide, 0.05MG sodium azide, 0.05MG thimerosal, 0.05MG thimerosal | |
| P97812, Q14623 | |
| Ihh | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human IhH. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| IHH | |
| Indian hedgehog protein, IHH, HHG-2, Indian hedgehog protein N-product, Indian hedgehog protein C-product, IHH | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16147, 3549, 84399 | |
| -20°C | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction