missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech IHCeasy WFS1 Ready-To-Use IHC Kit
Shop All Proteintech Products
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
Description
KHC0048 is a ready-to-use IHC kit for staining of WFS1. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you are steps away from obtaining high-quality IHC data.
Specifications
Specifications
| Product Type | Immunohistochemistry Kit |
| Target Species | Human, Mouse, Rat |
| Quantity | 50 Tests |
| Form | Mixture |
| Sample Type | FFPE Tissue |
| Kit Contents | 100 mL Antigen Retrieval Buffer, 2 x 100 mL Washing Buffer, 5 mL Blocking Buffer, 5 mL Primary Antibody, 5 mL Secondary Antibody, 0.2 mL Chromogen Component A, 4 mL Chromogen Component B, 5 mL Signal Enhancer, 5 mL Counter Staining reagent, 5 mL Mounting Media, Datasheet, Manual |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction