missing translation for 'onlineSavingsMsg'
Learn More

Proteintech IHCeasy Syntaxin 1A/1B Ready-To-Use IHC Kit

Product Code. 17738074 Shop All Proteintech Products
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
This item is not returnable. View return policy

Product Code. 17738074

Brand: Proteintech KHC005150T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A ready-to-use IHC kit for staining of Syntaxin 1A and Syntaxin 1B.

KHC0051 is a ready-to-use IHC kit for staining of Syntaxin 1A and Syntaxin 1B. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you are steps away from obtaining high-quality IHC data.

Specifications

Product Type Immunohistochemistry Kit
Target Species Human, Mouse, Rat
Quantity 50 Tests
Form Mixture
Sample Type FFPE Tissue
Kit Contents 100 mL Antigen Retrieval Buffer, 2 x 100 mL Washing Buffer, 5 mL Blocking Buffer, 5 mL Primary Antibody, 5 mL Secondary Antibody, 0.2 mL Chromogen Component A, 4 mL Chromogen Component B, 5 mL Signal Enhancer, 5 mL Counter Staining reagent, 5 mL Mounting Media, Datasheet, Manual
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.