Learn More
Proteintech IHCeasy SREBF2 Ready-To-Use IHC Kit
Product Code. 18056395 Shop All Proteintech Products
Click to view available options
Quantity:
50 Tests
Unit Size:
50 tests
Product Code. 18056395
Brand: Proteintech KHC168950TS
KHC1689 is a ready-to-use IHC kit for staining of SREBF2.
KHC1689 is a ready-to-use IHC kit for staining of SREBF2. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Quantity 50 Tests
Form Liquid