missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KHC0053 is a ready-to-use IHC kit for staining of smooth muscle actin (ASMA). The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you are steps away from obtaining high-quality IHC data.
Specifications
Specifications
| Product Type | Immunohistochemistry Kit |
| Target Species | Human, Mouse, Rat |
| Quantity | 50 Tests |
| Form | Mixture |
| Sample Type | FFPE Tissue |
| Kit Contents | 100 mL Antigen Retrieval Buffer, 2 x 100 mL Washing Buffer, 5 mL Blocking Buffer, 5 mL Primary Antibody, 5 mL Secondary Antibody, 0.2 mL Chromogen Component A, 4 mL Chromogen Component B, 5 mL Signal Enhancer, 5 mL Counter Staining reagent, 5 mL Mounting Media, Datasheet, Manual |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?