Learn More
Proteintech IHCeasy RXRG Ready-To-Use IHC Kit
Product Code. 18017385 Shop All Proteintech Products
Click to view available options
:
Quantity: 50 Tests
Product Code. 18017385
Brand: Proteintech KHC161150TS
KHC1611 is a ready-to-use IHC kit for staining of RXRG.
KHC1611 is a ready-to-use IHC kit for staining of RXRG. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Quantity 50 Tests
Form Liquid