missing translation for 'onlineSavingsMsg'
Learn More

Proteintech IHCeasy P62,SQSTM1 Ready-To-Use IHC Kit

Product Code. 17690117 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17690117 50 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17690117 Supplier Proteintech Supplier No. KHC005850T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ready-To-Use IHC Kit

KHC0058 is a ready-to-use IHC kit for staining of P62(SQSTM1). The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.

Specifications

Clonality Monoclonal
Target Species Human
Quantity 50 Tests
For Use With (Application) Immunohistochemistry (IHC)
Species Mouse
Content And Storage All the reagents are stored at 2°C to 8°C. The kit is stable for 6 months from the date of receipt.
Kit Contents
  • Antigen Retrieval Buffer(100 mL, 50 x)
  • Washing Buffer(100 mL x2, 20 x)
  • Blocking Buffer(5 mL, RTU)
  • Rabbit Polyclonal Primary Antibody(5 mL, RTU)
  • Polymer-HRP-Goat anti-Mouse Secondary Antibody(5 mL, RTU)
  • Chromogen Component A(0.2 mL, RTU)
  • Chromogen Component B(4 mL, RTU)
  • Signal Enhancer(5 mL, RTU)
  • Counter Staining Reagent(5 mL, RTU)
  • Mounting Media(5 mL, RTU)
  • Datasheet (1 Copy)
  • Manual(1 Copy)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.