Learn More
Proteintech IHCeasy NFE2 Ready-To-Use IHC Kit
Product Code. 18058935 Shop All Proteintech Products
Click to view available options
:
Quantity: 50 Tests
Product Code. 18058935
Brand: Proteintech KHC176150TS
KHC1761 is a ready-to-use IHC kit for staining of NFE2.
KHC1761 is a ready-to-use IHC kit for staining of NFE2. The kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use. Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Quantity 50 Tests
Form Liquid