missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- This kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use.
- Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Specifications
| Target Species | Human, Rat, Mouse |
| Sample Type | FFPE Tissue |
| Content And Storage | Store at 2°C to 8°C |
| Packaging Quantity | 50 Tests |
| Sufficient For | 50 Reactions |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?